Learn More
Biolegend Purified anti-p53
Supplier:  Biolegend 645701
Encompass_Preferred
Purified anti-p53 [DO-1]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: WB, ICC, IP; Size: 25 ug
Catalog No. 50-170-996