Learn More
Biolegend Purified anti-p63 (ΔN)
Supplier:  Biolegend 619001
Encompass_Preferred
Purified anti-p63 (ΔN) [Poly6190]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human (recognizes δN-isoforms); Apps: WB, IHC; Size: 50 uL
Catalog No. 50-606-021