Learn More
Biolegend Purified anti-p97/VCP
Supplier:  Biolegend 636801
Encompass_Preferred
Purified anti-p97/VCP [4G9]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, IP, ICC; Size: 25 ug
Catalog No. 50-170-914