Learn More
Biolegend Purified anti-PAK1
Supplier:  Biolegend 856701
Encompass_Preferred
Purified anti-PAK1 [A17137A]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: WB, IHC-P; Size: 25 ug
Catalog No. 50-207-4776