Learn More
Biolegend Purified anti-Pax6
Supplier:  Biolegend 862001
Encompass_Preferred
Purified anti-Pax6 [AD2.35]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: WB, IHC-P, ICC; Size: 25 ug
Catalog No. 50-207-4861