Learn More
Biolegend Purified anti-human/mouse/rat PCNA
Supplier:  Biolegend 307901
Encompass_Preferred
Purified anti-human/mouse/rat PCNA [PC10]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat, Cross-Reactivity: Other species; Apps: ICFC, ICC, IHC, IP, WB; Size: 25 ug
Catalog No. 50-166-445