Learn More
Biolegend Purified anti-Phosphotyrosine
Supplier:  Biolegend 309301
Encompass_Preferred
Purified anti-Phosphotyrosine [PY20]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse, Rat, All Species; Apps: WB, ICFC, ICC, IP; Size: 25 ug
Catalog No. 50-166-503