Learn More
Biolegend Purified anti-PLK-1
Supplier:  Biolegend 627702
Encompass_Preferred
Purified anti-PLK-1 [3F8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, KO/KD-WB, IP, ICC; Size: 100 ug
Catalog No. 50-170-797