Learn More
Biolegend Purified anti-PLK-1 Phospho (Thr210)
Supplier:  Biolegend 628901
Encompass_Preferred
Purified anti-PLK-1 Phospho (Thr210) [2A3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB; Size: 25 ug
Catalog No. 50-170-823