Learn More
Biolegend Purified anti-PSMA6
Supplier:  Biolegend 670602
Encompass_Preferred
Purified anti-PSMA6 [O91F3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB; Size: 100 ug
Catalog No. 50-208-7285