Learn More
Biolegend Purified anti-PYK2
Supplier:  Biolegend 685002
Encompass_Preferred
Purified anti-PYK2 [P85E2D5]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: WB; Size: 100 ug
Catalog No. 50-208-7499