Learn More
Biolegend Purified anti-rat CD11b/c
Supplier:  Biolegend 201801
Encompass_Preferred
Purified anti-rat CD11b/c [OX-42]; Isotype: Mouse IgG2a, κ; Reactivity: Rat; Apps: FC, IHC-F, IP; Size: 100 ug
Catalog No. 50-165-354