Learn More
Biolegend Purified anti-rat CD28
Supplier:  Biolegend 200902
Encompass_Preferred
Purified anti-rat CD28 [JJ319]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC, IP; Size: 500 ug
Catalog No. 50-165-319