Learn More
Biolegend Purified anti-rat CD3
Supplier:  Biolegend 201401
Encompass_Preferred
Purified anti-rat CD3 [1F4]; Isotype: Mouse IgM, κ; Reactivity: Rat; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-165-333