Learn More
Biolegend Purified anti-rat CD71
Supplier:  Biolegend 204401
Encompass_Preferred
Purified anti-rat CD71 [OX-26]; Isotype: Mouse IgG2a, κ; Reactivity: Rat; Apps: FC; Size: 100 ug
Catalog No. 50-207-2523