Learn More
Biolegend Purified anti-rat Ig light chain κ
Supplier:  Biolegend 407802
Encompass_Preferred
Purified anti-rat Ig light chain κ [MRK-81]; Isotype: Mouse IgG; Reactivity: Rat; Apps: ELISA, FC; Size: 500 ug
Catalog No. 50-169-212