Learn More
Biolegend Purified anti-RIG-1
Supplier:  Biolegend 635201
Encompass_Preferred
Purified anti-RIG-1 [SS1A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: WB; Size: 25 ug
Catalog No. 50-170-902