Learn More
Biolegend Purified anti-RORg
Supplier:  Biolegend 644902
Encompass_Preferred
Purified anti-RORg [4F3-3C8-2B7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Weakly Cross-Reactive with Mouse; Apps: WB; Size: 100 ug
Catalog No. 50-170-987