Learn More
Biolegend Purified anti-RUNX1
Supplier:  Biolegend 607801
Encompass_Preferred
Purified anti-RUNX1 [W16118A]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB; Size: 25 ug
Catalog No. 50-208-6651