Learn More
Biolegend Purified anti-Runx3
Supplier:  Biolegend 697902
Encompass_Preferred
Purified anti-Runx3 [R3-5G4]; Isotype: Mouse IgG1; Reactivity: Human; Apps: WB, IP; Size: 100 ug
Catalog No. 50-208-7708