Learn More
Biolegend Purified anti-SARS-CoV-2 S Protein S1 Recombinant
Supplier:  Biolegend 940701
Encompass_Preferred
Purified anti-SARS-CoV-2 S Protein S1 Recombinant [AM038105]; Isotype: Human IgG1, κ; Reactivity: SARS-CoV-2; Apps: ELISA Detection, ELISA; Size: 25 ug
Catalog No. 50-207-5420