Learn More
Biolegend Purified anti-SENP2
Supplier:  Biolegend 618401
Encompass_Preferred
Purified anti-SENP2 [W17126B]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: WB; Size: 25 ug
Catalog No. 50-208-6764