Learn More
Biolegend Purified anti-SHC1
Supplier:  Biolegend 669701
Encompass_Preferred
Purified anti-SHC1 [O91H8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, IP, ICC, IHC-P; Size: 25 ug
Catalog No. 50-208-7274