Learn More
Biolegend Purified anti-SIK2
Supplier:  Biolegend 636702
Encompass_Preferred
Purified anti-SIK2 [S15G10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IP; Size: 100 ug
Catalog No. 50-170-913