Learn More
Biolegend Purified anti-SIRT5
Supplier:  Biolegend 698801
Encompass_Preferred
Purified anti-SIRT5 [O91G9]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: WB, KO/KD-WB; Size: 25 ug
Catalog No. 50-208-7726