Learn More
Biolegend Purified anti-SKIL (SnoN)
Supplier:  Biolegend 644201
Encompass_Preferred
Purified anti-SKIL (SnoN) [O91H2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB; Size: 25 ug
Catalog No. 50-208-6904