Learn More
Biolegend Purified anti-SNAT1
Supplier:  Biolegend 857801
Encompass_Preferred
Purified anti-SNAT1 [N104/37]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: WB; Size: 25 ug
Catalog No. 50-207-4809