Learn More
Biolegend Purified anti-Spectrin β1
Supplier:  Biolegend 856902
Encompass_Preferred
Purified anti-Spectrin β1 [N385/21]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: WB; Size: 100 ug
Catalog No. 50-207-4782