Learn More
Biolegend Purified anti-STAT3
Supplier:  Biolegend 691502
Encompass_Preferred
Purified anti-STAT3 [P83B5]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB; Size: 100 ug
Catalog No. 50-208-7598