Learn More
Biolegend Purified anti-STAT3 Phospho (Ser727) [A16089A]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, ICC; Size: 25 ug
Supplier:  Biolegend 669501
Encompass_Preferred
Purified anti-STAT3 Phospho (Ser727) [A16089A]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, ICC; Size: 25 ug
Catalog No. 50-237-8572