Learn More
Biolegend Purified anti-STAT3 Phospho (Tyr705)
Supplier:  Biolegend 651001
Encompass_Preferred
Purified anti-STAT3 Phospho (Tyr705) [13A3-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB; Size: 25 ug
Catalog No. 50-171-084