Learn More
Biolegend Purified anti-Syk
Supplier:  Biolegend 646002
Encompass_Preferred
Purified anti-Syk [5F5]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: WB, IP; Size: 100 ug
Catalog No. 50-171-003