Learn More
Biolegend Purified anti-Syk
Supplier:  Biolegend 644302
Encompass_Preferred
Purified anti-Syk [4D10.2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: WB, IP, ICFC; Size: 100 ug
Catalog No. 50-170-965