Learn More
Biolegend Purified anti-Syk
Supplier:  Biolegend 644301
Encompass_Preferred
Purified anti-Syk [4D10.2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: WB, IP, ICFC; Size: 25 ug
Catalog No. 50-170-964