Learn More
Biolegend Purified anti-Tau, 359-373
Supplier:  Biolegend 851401
Encompass_Preferred
Purified anti-Tau, 359-373 [A16097B]; Isotype: Rat IgG2b, κ; Reactivity: Human, Mouse, Rat; Apps: WB, Direct ELISA, IHC-P; Size: 25 ug
Catalog No. 50-207-4642