Learn More
Biolegend Purified anti-Tau, 368-441
Supplier:  Biolegend 851201
Encompass_Preferred
Purified anti-Tau, 368-441 [A16097F]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB, Direct ELISA, IHC-P; Size: 25 ug
Catalog No. 50-207-4632