Learn More
Biolegend Purified anti-Tau, 425-441
Supplier:  Biolegend 851102
Encompass_Preferred
Purified anti-Tau, 425-441 [A16097E]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: WB, IHC-P; Size: 100 ug
Catalog No. 50-207-4629