Learn More
Biolegend Purified anti-Tau Phospho (Thr181)
Supplier:  Biolegend 846601
Encompass_Preferred
Purified anti-Tau Phospho (Thr181) [M7004D06]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: IHC-P, Direct ELISA, WB; Size: 25 ug
Catalog No. 50-207-4499