Learn More
Biolegend Purified anti-Thioredoxin 1
Supplier:  Biolegend 658901
Encompass_Preferred
Purified anti-Thioredoxin 1 [4H12A59]; Isotype: Mouse IgG1, κ; Reactivity: E. Coli Thioredoxin-1 and thioredoxin tagged recombinant protein; Apps: WB, IP; Size: 25 ug
Catalog No. 50-208-7156