Learn More
Biolegend Purified anti-TIF1β (KAP-1, TRIM28)
Supplier:  Biolegend 619301
Encompass_Preferred
Purified anti-TIF1β (KAP-1, TRIM28) [20A1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, ICC, ChIP; Size: 25 ug
Catalog No. 50-170-713