Learn More
Biolegend Purified anti-TRAF3
Supplier:  Biolegend 680802
Encompass_Preferred
Purified anti-TRAF3 [9D7C15]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB; Size: 100 ug
Catalog No. 50-208-7419