Learn More
Biolegend Purified anti-Transthyretin
Supplier:  Biolegend 856301
Encompass_Preferred
Purified anti-Transthyretin [A16143A]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: WB, IHC-P; Size: 25 ug
Catalog No. 50-207-4762