Learn More
Biolegend Purified anti-Transthyretin, aggregated
Supplier:  Biolegend 848101
Encompass_Preferred
Purified anti-Transthyretin, aggregated [TA5F4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Capture, IHC-P, Direct ELISA; Size: 25 ug
Catalog No. 50-207-4531