Learn More
Biolegend Purified anti-Tubulin-α
Supplier:  Biolegend 627902
Encompass_Preferred
Purified anti-Tubulin-α [10D8]; Isotype: Mouse IgM, κ; Reactivity: Human, Mouse, Rat, All Species; Apps: WB, ICC; Size: 100 ug
Catalog No. 50-170-803