Learn More
Biolegend Purified anti-Tubulin β 3 (TUBB3)
Supplier:  Biolegend 801202
Encompass_Preferred
Purified anti-Tubulin β 3 (TUBB3) [TUJ1]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB, ICC, FC; Size: 250 uL
Catalog No. 50-102-9509