Learn More
Biolegend Purified anti-Tubulin Beta 3 (TUBB3)
Supplier:  Biolegend 657402
Encompass_Preferred
Purified anti-Tubulin Beta 3 (TUBB3) [AA10]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: WB, ICC; Size: 100 ug
Catalog No. 50-605-928