Learn More
Biolegend Purified (azide-free) anti-Tyrosine Hydroxylase
Supplier:  Biolegend 818001
Encompass_Preferred
Purified (azide-free) anti-Tyrosine Hydroxylase [2/40/15]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB; Size: 100 uL
Catalog No. 50-102-9771