Learn More
Biolegend Purified anti-UQCRC1
Supplier:  Biolegend 697602
Encompass_Preferred
Purified anti-UQCRC1 [O91G6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, KO/KD-WB. ICC; Size: 100 ug
Catalog No. 50-208-7702