Learn More
Biolegend Purified anti-VASP
Supplier:  Biolegend 937501
Encompass_Preferred
Purified anti-VASP [W18344B]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: WB, IP, ICC; Size: 25 ug
Catalog No. 50-207-5372