Learn More
Biolegend Purified anti-XBP-1s
Supplier:  Biolegend 658802
Encompass_Preferred
Purified anti-XBP-1s [9D11A43]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse, Human; Apps: WB, ICC, ChIP; Size: 100 ug
Catalog No. 50-605-943